Air Purifying Dual Cartridge Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
54001 54001S 760008AS 770030S 770030M 770030L 760008A 550030S 550030M 550030M
CA45000-754EA 336.03 CAD
CA45000-754 CA45000-756 CA56222-952 CA56222-944 CA56222-946 CA56222-948 CA56222-950 CA56223-220 CA56223-222 CA56223-224 56223-222
Air Purifying Dual Cartridge Respirators, Honeywell Safety
Respirators Reusable Respirators Full Face Respirators
Comfortable half-mask or full facepiece respirators for use with North Safety Products cartridges and filters.
  • Reliable and latex-free respirators offer direct cartridge-to-facepiece connection
  • 7700 and 7600 series have nonallergenic silicone facepieces
  • 5500 and 5400 series have thermoplastic elastomer facepieces
  • NIOSH approved when used with North Safety Products cartridges
Interchangeable cartridges and filters provide air purifying protection for a variety of applications.
Half-mask respirators fit snugly while the cradle suspension systemMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable