VACUUBRAND® General-Purpose Oil-Free Diaphragm Vacuum Pumps, BrandTech
Supplier: Brandtech
20731000 20731003 20734000 20734003 20732000 20732003 20732200 20736000 20736003 20738000 20738003 20730003
97007-150EA 7624.91 CAD
97007-150 97007-152 97007-154 97007-156 97007-158 97007-160 97007-162 97007-166 97007-168 97007-170 97007-172 89202-292 89511-182
VACUUBRAND® General-Purpose Oil-Free Diaphragm Vacuum Pumps, BrandTech
Pumps Vacuum Pumps
Designed specifically for non-corrosive and non-evaporative procedures, oil-free diaphragm vacuum pumps provide quiet and stable continuous-duty performance
  • For non-evaporative, non-corrosive applications
  • Integrated silencer(s)
  • High performance at working vacuum
  • 1-, 2- 3- and 4- stage designs
  • MD 4 and MZ 2D models feature KF16 flange inlets
  • All others have hose barbs
These aluminum-FKM diaphragm pumps utilize dry operation technology, eliminating the need to change or monitor oil. Diaphragms provide 10,000 to 15,000 hours of use before replacement, minimizing downtime and servicing. The unique pump design eliminates tedious, trial and error stroke length recalibration. Individually accessible diaphragms and valves simplify servicing. All pumps are equipped with specially engineered pump heads for high flow rates at working vacuum, allowing for reduced process times and higher flow throughput. Before shipping, all pumps must pass rigorous product testing to ensure reliability.
Ordering information: Pumps are supplied complete with one phase motor, on/off switch, cable with plug, operating manual, and a one-year manufacturer's warranty. The 230V models include a European-style plug. MD1 Diaphragm Pump model 20696087 is a dual-voltage model, and is not supplied with an IEC/mains cord.