VWR® & VWR® Signature™ Multi-Tube Vortexers
Supplier: VWR International
14005-826 58816-115 10027-190
14005-826EA 9550.91 CAD
14005-826 58816-115 10027-190
VWR® & VWR® Signature™ Multi-Tube Vortexers
Shakers Vortex Mixers
These vortexers are designed to hold multiple test tubes, vials, or cylinders.
  • Process up to 50 samples at a time
  • Continuous or timed operation
  • Includes foam rack for 12 mm tubes
  • Now with best-in-class, exclusive 5-year warranty
The vortexing action is created by securing the top of the vessel in place while allowing the bottom to move freely in a defined 3.6 mm eccentric orbit.
VWR® Signature™ digital vortexers feature a microprocessor controller for digital programming of both speed and time, along with a pulse feature for difficult mixingMore Product Information