8210 Disposable N95 Respirator, 3M™
Supplier: 3M
8210 8210
CA56222-362CS 366.91 CAD
56222-362 CA56222-362
8210 Disposable N95 Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
Economical respirator provides reliable worker protection against certain solid and liquid aerosols that do not contain oil.
  • Provides reliable worker protection against certain solid and liquid aerosols that do not contain oil
  • Provides minimum filtration efficiency of 95%
  • Lightweight construction for greater worker comfort
  • Adjustable nose clip for custom fit and secure seal
Provides minimum filtration efficiency of 95%. Lightweight construction for greater worker comfort. Adjustable noseclip for custom fit and secure seal.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.