VACUUBRAND® Dual Application Dry Chemistry Vacuum Systems, BrandTech
Supplier: Brandtech
20732800 20732803 20736800 20736803
97007-190EA 17499.56 CAD
97007-190 97007-192 97007-198 97007-200 97007-214 97007-216 97007-218 97007-220 97007-226 97007-228 97007-230 97007-232
VACUUBRAND® Dual Application Dry Chemistry Vacuum Systems, BrandTech
Pumps Vacuum Pumps
Dual application dry chemistry vacuum systems feature reliable and powerful pumps to provide whisper-quiet vacuum operation with integrated solvent recovery accessories
  • Whisper-quiet
  • Integrated flow control for two applications
  • Check-valves minimize interaction
  • Typical service interval >15,000 operating hours
  • Energy-efficient
  • Space-saving design
Pump systems are engineered for high-performance operation, with flow rates up to 40% higher than competitive models at working vacuum levels. The chemistry-design diaphragm vacuum pumps feature flowpathsMore Product Information