2300 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
2300N95 2307N95
56219-020CS 1347.69 CAD
56219-020 56219-025
2300 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Disposable respirators provide a filter efficiency level of 95% or greater against oil-free particulate aerosols up to 10 times OSHA Permissible Exposure Limit (PEL).
  • Disposable
  • Crush-resistant Dura-Mesh® shell
  • Filter efficiency level of 95% or greater against oil-free particulate aerosols
  • Up to 10X OSHA Permissible Exposure Limit (PEL)
Exhale valve lets worker breathe out easily, keep cool longer
Medium/large size respirators are also available in a locker/dispenser for easy storage and dispensing. Locker/dispenser contains a personal supply of five respirators. A separate locker compartment allows storage of a used respirator during worker breaks. Helps prevent premature disposal.
The contoured facepiece with Softspun® lining keeps worker comfortable. Durable mask with crush-resistant Dura-Mesh® outer shell will not absorb moisture or collapse. Stays clean-looking longer for more wear, greater economy. Straps are securely attached so they do not break at the seal. Available in three sizes: small, medium/large, and alternate shape (for workers with rounder facial features and flatter nose bridges).