8247 R95 Respirator, 3M™
Supplier: 3M
7100160544 7000002060
CA11006-300CS 907.14 CAD
CA11006-300 CA56621-932
8247 R95 Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
This respirator provides worker protection against certain oil and non-oil based aerosols including those with nuisance levels of organic vapors such as solvents, degreasers, and resins.
  • Protects against certain oil and non-oil based aerosols
  • If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use
  • Provides minimum filtration efficiency of 95%
(Nuisance level acid gas refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) If oil aerosolsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable