KIMTECH PURE® M5 Face Masks, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
62692 62742 62692
CA10815-674CS 239.28 CAD
CA10815-674 CA10869-020 10815-674
KIMTECH PURE® M5 Face Masks, Kimberly-Clark Professional®
Masks Cleanroom Masks
These non sterile pleat-style masks filter particles and bacteria.
Recommended for ISO Class 5 or higher cleanroom environments.
Masks feature a three-layer design with polyester/cellulose inner/outer facing.
Color: blue.
Packaging: Double-bagged.