Synthware Buchner Filter Funnels, Inner [ST] Joint, Kemtech America
Supplier: Kemtech America
F362014C F362014M F362019C F362019M F362024C F362024M F363014C F363014M F363019C F363019M F363024C F363024M F364014C F364014M F364019C F364019M F364024C F364024M F366024C F366024M F366029C F366029M F368024C F368024M F369024C F369024M
60003-444EA 147.95 CAD
60003-444 60003-446 60003-448 60003-450 60003-452 60003-454 60003-456 60003-458 60003-460 60003-462 60003-464 60003-466 60003-468 60003-470 60003-472 60003-474 60003-476 60003-478 60003-484 60003-486 60003-488 60003-490 60003-492 60003-494 60003-500 60003-502
Synthware Buchner Filter Funnels, Inner [ST] Joint, Kemtech America
Funnels Filtration Funnels Büchner Funnels
These filter funnels are made of glass.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Medium and coarse frit porosity available
  • Barbed side arm
Note: Not recommended for filtration where pressure would exceed 15 psi.