Draft Chamber Kits, METTLER TOLEDO®
Supplier: Mettler Toledo
63052324 63052337
CA11277-752EA 975.36 CAD
CA11277-752 CA11277-985
Draft Chamber Kits, METTLER TOLEDO®
Shields Safety Shields
To help eliminate drafts and stabilize balance readings, these 4.8 mm thick Plexiglas® chambers create a draft-free area around your METTLER TOLEDO® balance.
  • Eliminate drafts
  • Stabilize balance readings
Ordering information: Assembly required.