Tubing, Tubing Cartridges, and Accessories for Peristaltic Pumps, Welch®, Gardner Denver
Supplier: Welch Vacuum
3432K-02 3434K-01 3434K-02 3434K-04 3232K-01 3232K-02 3234K-02 3234K-04
84009-526PK 146.02 CAD
84009-526 84009-536 84009-538 84009-540 84009-544 84009-546 84009-558 84009-560 84009-704
Tubing, Tubing Cartridges, and Accessories for Peristaltic Pumps, Welch®, Gardner Denver
Tubing Transfer Tubing
For use with Welch® peristaltic pumps (see 84009-696 series).
Silicone tubing is flexible and translucent with good chemical and environmental resistance and very good electrical resistance. High-performance, flexible PVC tubing features great versatility and moderate chemical resistance and is ideal for use with peristaltic pump heads. Lok-n-Flow tubing cartridges allow easy replacement of worn tubing at the pump head without wasted materials.
Ordering information: The starter tubing kit 84009-704 includes 7.6m (25') of 4.8mm I.D. general PVC tubing; four hose clamps; and two each of 1More Product Information