Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
BS07A-RD BS07A-BU BS07A-YW BS08A-RD BS08A-BU BS08A-GN BS08A-YW
CA82004-216EA 37.23 CAD
CA82004-216 CA82004-218 CA82004-220 CA82004-222 CA82004-224 CA82004-226 CA82004-228
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Security Locks
Designed to lock a wide range of valves, these small and large ball valve lockouts are both versatile and easy to use
Two halves encompass the ball valve handle to secure it from inadvertent use. Lockouts are made from ultra-tough polypropylene.