Safety Padlock, Brady®
Supplier: Brady Worldwide
51339 51344 51345 51346 51347 51353
CA89007-292PK 122.41 CAD
CA89007-292 CA89007-294 CA89007-296 CA89007-298 CA89007-300 CA89007-302
Safety Padlock, Brady®
Security Locks
Brady® Safety Padlocks are the ultimate locks for a wide variety of safety and security needs.
  • Unique cylinder is insulated to protect workers from shocks
  • Tamper resistant
  • Ribbed design for easier gripping and handling
Unique cylinder is insulated to protect workers from shocks when key is inserted. They cylinder is 6-pin precision-machined for more unique key cuts and better tamper resistance. Reserved, paracentric keyway provides optimal security and the special design prohibits the key from being released until the padlock shackle is closed. Locks are compact and lightweight, and they resist impact, temperature, chemicals, and corrosion. Their nonconductive, nonsparking body features a ribbed design for easier gripping and handling.
Shackle clearance is 38.1mm (1½"), shackle diameter is 6.35mm (¼"). Lock body measures 44.45H x 38.1W x 20.32D mm (1¾ x 1½ x ⅘").
Ordering information: Locks come standard with one key and English, French, and Spanish language labels. Padlocks are available keyed differently. Custom keying and charting is available.