Lindberg/Blue M Vacuum Ovens, Thermo Scientific
Supplier: Thermo Fisher Scientific
VO914A-1 VO1218A-1
52346-046EA 41471.98 CAD
52346-046 52346-050 52348-350 52348-360
Lindberg/Blue M Vacuum Ovens, Thermo Scientific
Ovens Vacuum Ovens
Vacuum ovens are designed for drying, curing, outgassing, aging, process control, and other applications which require elevated temperatures in reduced atmospheres, or vacuum/purge with nonflammable and inert atmospheres
Fiberglass insulation minimizes heat loss in walls and door. Stainless steel interiors provide resistance against corrosive materials. Oven exteriors are available in stainless steel or white painted finish. Ovens with stainless steel exteriors feature solid outer door with inner window door.
Caution: Not suitable for use with hazardous vapors.
All ovens featureMore Product Information