Drum Truck, Wesco
Supplier: Wesco
240005 240006
21002-004EA 1043.29 CAD
21002-004 21002-008
Drum Truck, Wesco
Trolleys
Four-wheeled, multi-functional drum truck allows handling of 208.2 L (55 gal.) steel drum and acts as a drum cradle for storage or dispensing
Non-sparking polyolefin rollers help rotate drum when truck is placed in a horizontal (cradle) position. Rear axle allows loading/unloading of drums from pallets. Use handy kickstand for stable upright storage.
Ordering information: Available with rubber rimmed cast iron wheels or polyolefin wheels (economical polyolefin resists most environments that may be damaged by rubber wheels).