Orbitron Rotators, Boekel Scientific
Supplier: Boekel
260250 260200
35821-000EA 1749.66 CAD
35821-000 56261-508
Orbitron Rotators, Boekel Scientific
Shakers Rotating/Rocking Shakers
Gentle, uniform, low-foam mixing action suitable for immunoassays, hybridizations, mixing blood samples, coupling affinity chromatography media, and DNA extractions
Rotator I features a fixed speed of 20rpm and tilt of 23°. Rotator II features variable speed of 6–23rpm and tilt of 0–23°.
Compact unit has 30.5cm (12") square platform. Dimpled pad on platform accepts a variety of sizes of tubes simultaneously with no need for racks or clamps. Pad is removable for easy cleaning and test tube transport.
Ordering information: Rotator I is supplied with one dimpled pad. Rotator II isMore Product Information