VWR® Hybridization Ovens
Supplier: VWR International
47746-130 97005-254 97005-252
47746-130EA 3818.01 CAD
47746-130 97005-254 97005-252
VWR® Hybridization Ovens
Ovens Hybridisation Ovens
Hybridization Ovens are designed to provide rapid heat-up, excellent temperature stability, and efficient mixing for all types of hybridization and incubation applications.
  • Compact design
  • Smooth, quiet, and durable
  • VWR® Two-Year Limited Parts and Labor Warranty
Model 5430, a space-saving mini unit, holds up to four 50 mL conical tubes
Optional multi-tube carousel (47746-116) includes clips to hold twelve 1.5 mL or 2.0 mL conical tubes, six 15 mL conical tubes, or four 50 mL conical tubes. Unit heats from ambient to 70°C in 25 minutes. Models 230502V and 230502V-2 are medium-capacity units, holding up to four 35x300mm, or four 35x225mm, or eight 35x150mm, or eight 35x100mm bottles. Unit heats from ambient to 65°C in 20 minutes.
Optional multi-tube carousel (47746-114) holds forty 1.5 mL or 2.0 mL conical tubes, twelve 15 mL conical tubes, or ten 50 mL conical tubes.
Model 5420 and 5420-2 are high-capacity units, holding up to ten 35x300mm bottles or twenty 35x150mm bottles. Optional large bottle carousel (47746-110) holds four 70x220mm (750 mL) bottles or four 55x180mm (400 mL) bottles. Optional multi-tube carousel (47746-112) holds one hundred 1.5 mL or 2.0 mL conical tubes, thirty-five 15 mL conical tubes, or thirty 50 mL conical tubes. Optional microplate carousel (47746-118) holds 10 sealed standard microplates or deep well plates. Unit heats from ambient to 65°C in 30 minutes.
Note: Model 5430 oven is not recommended for use with polystyrene tubes. Plates must be sealed before use in microplate carousels.
Ordering information: Model 5420 includes two standard 35x300mm glass hybridization bottles, two spare O-rings, and a stainless steel drip pan. For Models 230502V and 5420 optional multi-tube carousels, please order clips separately. Glass bottles are supplied with blue caps and silicone O-rings for standard procedures or red caps and PTFE seals for high temperature procedures. Acrylic bottles are constructed of beta-blocking acrylic and are supplied with caps and leakproof gaskets. For 230V capability, contact your VWR sales representative.