Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Supplier: Thermo Fisher Scientific
BF51842C-1 BF51842PBC-1 BF51731BC-1
30604-200EA 19592 CAD
30604-200 30604-203 30712-122 30712-132
Lindberg/Blue M Box Furnaces, 1200 °C, Thermo Scientific
Ovens Furnaces (Ashing and Muffle Ovens)
Furnaces are designed for continuous operation to 1200 °C maximum and feature an integral controller in a single space-saving cabinet.
  • LGO heating elements and Moldatherm insulation
  • Variable heatup rate eliminates thermal shock to materials
  • Air vent (1" diameter, top) and air inlet (0.375" diameter, rear) for inert gas
  • Platinel II thermocouple for long-term stability
  • Removable shelves for versatility
  • Self-tuning PID control provides optimum thermal process
  • Adjustable high-limit OTP
  • LED display of actual temperature vs. setpoint
Light gauge overbend (LGO™) heating element embedded within Moldatherm® ceramic fiber insulation provides optimum temperature uniformity, energy efficiency, fast response, and rapid heat-up and cool-down. Microprocessor-based self-tuning PID instrumentation automatically sets the best control parameter values during operation. Safety door switch interrupts power to heating element when door is opened. A 2.54 cm (1") diameter air vent on top and 9.5mm (3/8") diameter air inlet on rear of furnace allow for inert atmosphere exchange.
Single setpoint units provide a single segment. Single programmable units have 16 segments and 1 program for ramps and dwells. Multiple programmable controllers feature 300 segments with 30 programs for ramps and dwells. All units feature an adjustable over-temperature protection and LED readout to display actual temperature vs. setpoint in °C or °F. Units are also available with optional over-temperature control that shuts unit off automatically if high limit is reached.
Choose from vertical lift door or horizontal side swing door. An optional adjustable gas flowmeter, located on the front panel, is available on 300-segment controllers to allow fresh air exchange for ashing applications or use with inert atmosphere. Chamber is not air-tight.
Ordering information: Furnaces feature a one-year parts and labor warranty. Platinel® II thermocouple and Moldatherm® hearth plate included. Power cord and hard wiring not included. Optional RS-485 data connection port is available on all models; contact your Avantor sales representative.