Parker® PROPOR BR DEMI Capsules
Supplier: PARKER HANNIFIN TE
ZEBRB-020TT-PN3 ZEBRB-020TT-PS3 ZEBRE-020GG-PN3 ZEBRE-020GG-PS3 ZEBRE-020HH-PN3 ZEBRE-020HH-PS3 ZEBRE-020TT-PN3 ZEBRE-020TT-PS3 ZEBRA-020GG-PN3 ZEBRA-020GG-PS3 ZEBRA-020HH-PN3 ZEBRA-020HH-PS3 ZEBRA-020TT-PN3 ZEBRA-020TT-PS3 ZEBRB-020GG-PN3 ZEBRB-020GG-PS3 ZEBRB-020HH-PN3 ZEBRB-020HH-PS3
77674-924EA 1009.45 CAD
77674-924 77674-942 77674-938 77674-956 77674-932 77674-950 77674-926 77674-944 77674-934 77674-952 77674-928 77674-946 77674-922 77674-940 77674-936 77674-954 77674-930 77674-948
Parker® PROPOR BR DEMI Capsules
Filters Membrane Filter Capsules
PROPOR BR filters have been specifically designed for fast and cost-effective bioburden reduction of pharmaceutical solutions.
  • Brevundimonas diminuta retention of LRV >5 for efficient bioburden reduction
  • Additional prefilter layer gives excellent throughput to blockage
  • Low binding for minimal product loss
PROPOR BR filters feature an integral meltblown prefilter layer to maximize dirt holding capacity whilst the polyethersulphone membrane guarantees a bioburden log reduction of greater than 5, giving excellent microbial protection. This makesMore Product Information