Rodent Rectal Temperature Probes
Supplier: WORLD PRECISION INSTRUMENTS LLC
RET-2 RET-3
77665-122EA 211.72 CAD
77665-122 77665-124
Rodent Rectal Temperature Probes
Temperature Probes
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.
  • Works with the ATC2000 TC input
  • Not isolated
  • 125 °C Max. Temperature
  • 5 ft. lead length
When precise temperature measurements are required, WPI can provide you with a very accurate monitor and thermocouple microprobes. WPI monitors have both resolution and accuracy of 0.1 °C in the 0 to 50 °C range and are traceable to NIST standards, whereas, other competitive electronic thermometers have an accuracy that is usually to 0.5 °C or worse. Furthermore, all our type T clinical probes are guaranteed accurate to 0.1 °C, due to our stringent wire standards. These are five times more accurate than competitive probes made with regular 'Special Limits' wire.
The needle microprobes and animal rectal probes are all designed to withstand sterilization by ETO gas and gluteraldehyde solution. The flexible implantable microprobes are both ETO gas sterilizable and autoclavable.