PROPOR MR Cartridges
Supplier: PARKER HANNIFIN TE
ZCMRK-610C-PE ZCMR1-610C-PE ZCMR2-610C-PE ZCMR3-610C-PE ZCMRK-610C-PS ZCMR1-610C-PS ZCMR2-610C-PS ZCMR3-610C-PS
76498-036EA 1351.18 CAD
76498-036 76498-038 76498-040 76498-042 76498-044 76498-046 76498-048 76498-050
PROPOR MR Cartridges
Filters Filter Cartridges
PROPOR MR filters have been specifically designed for the fast, effective, and economical removal of mycoplasma from cell culture media in the biopharmaceutical industry.
  • Fully validated and integrity testable for assurance of sterility
  • A typical LRV of >10 for Acholeplasma laidlawii for effective mycoplasma control
  • Integral prefilter layer increases throughputs for reduction of filter trains and exceptional flow rates for quick processing of cell culture media
Incorporating a highly retentive 0.1 micron rated PES membrane, PROPOR MR is validatedMore Product Information