MICRO-ePORE™ Electrode Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
300683PT 300684 300684PT 300685
77662-056EA 505.6 CAD
77662-056 77662-058 77662-060 77662-062
MICRO-ePORE™ Electrode Holders
Electrode Holders
These electrode holders are designed exclusively for use the with MICRO-ePORE™ system, which includes the silver wire electrode holders. The platinum wire electrode holders are an optional accessory.
Platinum wire electrode holders are important when you are working with proteins. The silver chloride wire is perfect for work with RNA. However, the silver chloride wire tends to denature proteins, reducing the efficiency of the microinjection solution.
Applications for silver wire electrode holders: Silver wire electrodes are suitable for most applications
Applications for platinum wire electrode holders: Platinum electrodes offers more active insertion of the genetic material capability, and there is a better chance of changing the genome