Corneoscleral Punches
Supplier: WORLD PRECISION INSTRUMENTS LLC
500079 500143
77662-124EA 274.95 CAD
77662-124 77662-136
Corneoscleral Punches
Animal Supplies Animal ID Tags and Punches
This corneoscleral punch is an excellent tool for opening the skull of a small animal without damaging the brain or spinal cord.
  • 8 cm (3") Long
  • Stainless steel
The lower jaw of the punch is a flat plate which can be inserted into the skull with much less damage to the brain than a rongeur.