Ring Handled and Angled Operating Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
500047-G 500047
77662-116EA 225.94 CAD
77662-116 77662-114
Ring Handled and Angled Operating Scissors
Scissors
Ring-handled and angled operating scissors are a pair of surgical scissors commonly used in general surgery for cutting and dissecting tissues.
  • Angled scissors for greater flexibility and control in confined spaces
  • Sharp/sharp tips
The 14 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. The angle of the blades allows for greater flexibility and precision in cutting inMore Product Information