Ring handled ear punches are commonly used in a laboratory setting for tagging rodents for easy identification. Ear punches can create a distinctive hole or pattern in the ear of a rodent.
- Non-invasive ear tagging for rodents
- Permanently mark your rodents
- Maintenance-free marking of rodents
- Inexpensive when compared with tattooing, ear tags, and RFID
- 10 cm (4") Long
- Choose 1 or 2 mm hole punch
- Reusable
- Sterilizable
- Surgical grade stainless steel
Ear punches are relatively non-invasive compared with ear tags, tattoos and other methods. They create a hole in the ear without causing significant harm or discomfort to the animal. Ear punches permanent mark your rodents, without ever fading or require further maintenance. You can easily identify individual animals throughout their lifespan.
The distinctive pattern of the ear punch can be easily seen and identified by laboratory personnel. Ear punches are relatively inexpensive compared to other tagging methods. Ring Handled Ear Punches offer several advantages over simple ear punches. The ring handles give you greater control over the punching action, which means you get more consistent results and fewer misalignments.
Reduce hand fatigue with ring handled ear punches, because they are more comfortable when you are tagging many animals. The ring handles also improve your visibility when you are tagging rodents. Our ring handled ear punches are reusable, reducing ongoing costs associated with tagging rodents.
These ear punches are manufactured from surgical grade stainless steel and are suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.