Mouse Recombinant FGF-Basic (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-12AF-100UG
200-12AF-10UG
200-12AF-1MG
200-12AF-500UG
200-12AF-50UG
77660-822EA
584.24
CAD
77660-822
77660-824
77660-826
77660-828
77660-830
Mouse Recombinant FGF-Basic (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only. Not for human or veterinary use.