Sterile Disposable Rinse Station Inserts of EVOM™ Auto
Supplier: WORLD PRECISION INSTRUMENTS LLC
EVA-AC-03-02-10 EVA-AC-03-02-25
77664-268PK 55.3 CAD
77664-268 77664-270
Sterile Disposable Rinse Station Inserts of EVOM™ Auto
First Aid Supplies Eyewash Stations and Eyewash Bottles
These disposable rinse stations fit conveniently inside the rinse station of the EVOM™ Auto Automated TEER Measurement system for effective and convenient electrode cleaning, disinfection, and maintenance. These are available in packages of 10 and 25.
  • Individually sterile rinse station inserts
  • Dispose after use
  • Use a fresh sterile one after each sample plate reading or before switching to different set of sample plates
  • Recommended for convenient and effective electrode cleaning, disinfection and maintenance