Frameworks
Supplier: WORLD PRECISION INSTRUMENTS LLC
503204 503205 503207 503208
77663-128EA 722.63 CAD
77663-128 77663-130 77663-132 77663-134
Frameworks
Furniture Tables and Workbenches
These high quality bases are made of stainless steel and polymer that resists organic solvents and corrosion. They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs. Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.
Frameworks starter kit (77663-128) kit includes 1× open-sided frame clamp (pack of four), 1× heavy rectangular base with M8 thread mount and thumbscrew mount, 23×15.6 cm, 4 lbs., 1× parallel frame clamp (pack of 4), 1× medium clamp with rod (157 mm), opens to 85 mm, 1× polished stainless steel post, ext.Ø 12 mm, 50 cm long, no thread, 1× polished stainless steel post, ext.Ø 12 mm, 60 cm long, M8 thread, 1× small clamp with rod (157 mm), opens up to 16 mm.
Frameworks small assembly (77663-130) kit includes 1× open-sided frame clamp (pack of four), 1× polished stainless steel post, ext.Ø 12 mm, 25 cm long, M8 thread, 1× light rectangular base with M8 thread mount and thumbscrew mount, 23×15.6 cm, 4 lbs., 1× small clamps with rod (157 mm), opens up to 16 mm.
Frameworks v-assembly small (77663-132) kit includes 1× open-sided frame clamp (pack of four), 1× polished stainless steel post, ext.Ø 12 mm, 25 cm long, M8 thread, 1× small clamps with rod (157 mm), opens up to 16 mm, 1× small V-base with M8 thread mount.
Frameworks v-assembly large (77663-134) kit includes 1× open-sided frame clamp (pack of four), 1× large clamp with rod (157 mm), opens to 85 mm, 1× polished stainless steel post, ext.Ø 12 mm, 75 cm long, M8 thread, 1× large V-base with M8 thread mount.