High Pressure Single-Syringe Pumps, Aladdin

Supplier: WORLD PRECISION INSTRUMENTS LLC
AL-1000HP AL-1000HP2
77664-046EA 1461.5 CAD
77664-046 77664-048
High Pressure Single-Syringe Pumps, Aladdin
Pumps Syringe Pumps
Aladdin High Pressure Single Syringe Pump is often used in research where precise, controlled delivery of fluids at elevated pressures is required. High-pressure syringe pumps can be used in chemical synthesis processes requiring precise control over the addition of reagents. This is especially important in reactions that occur under high pressures, such as certain types of catalytic reactions or polymerization processes.

  • Holds one Syringe up to 140 ml
  • Infusion rates from 1.733 µl/hr (1 ml syringe) to 340.6 ml/min (140 ml syringe)
  • Easy-to-use keypad interface
  • Space Saving Chassis: Foot print size of only 11 1/4×6 1/8"
  • Won't take up unnecessary space on your laboratory or production bench
  • Includes all the advanced functions and programming features of the Aladdin series

These high pressure Aladdin pumps hold a single syringe up to 140 ml (a 200 ml syringe partly filled). They can infuse from 1.733 µl/hr with a 1 ml syringe up to 340.6 ml/min with a 140 ml syringe. With all the advanced function and programming features of the Aladdin series of syringe pumps, you can choose between 100 lbs. (AL-1000HP) and 200 lbs.(AL-10000HP2) force.

High-pressure syringe pumps can also be utilized in biological and biochemical research for controlled delivery of solutions, such as high-pressure cell lysis, protein folding studies, or studies involving high-pressure biophysical techniques. Researchers studying catalysis, especially those focusing on heterogeneous catalysis, may use high-pressure syringe pumps to deliver reactants at controlled rates and pressures. This is important for understanding and optimizing catalytic processes. In microfluidics research, where small volumes of fluids are manipulated, high-pressure syringe pumps can be used to precisely control the flow of fluids through microchannels and devices.

Motor type: Step Motor, ¹/⁸ to ¹/₂ step modes
200 steps per revolutions
0.425 µm (¹/⁸ stepping) to 1.701 µm (¹/₂ stepping)
15/28 motor to drive screw ratio
Speed (max./min.): 18.4 cm/min/0.0084 cm/hr
Pumping rates: 6120 ml/hr with 60 ml syringe, to 1.459 µl/hr with 1 ml syringe
Maximum force: 100 lbs. at min. speed, 18 lbs. at max. speed
41 Number of program phases
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR