Single-Syringe Infusion Pumps, Aladdin
Supplier: WORLD PRECISION INSTRUMENTS LLC
AL-1000 AL-2000
77664-044EA 1287.7 CAD
77664-044 77664-050
Single-Syringe Infusion Pumps, Aladdin
Pumps Syringe Pumps
The AL-1000 and AL-2000 both offer the withdraw and inject functionality, as well as manually triggered injections. Both options have the capability to be programmed in multi-step pumping sequences without the requirement for a computer. Additionally, the AL-1000 and AL-2000 has both RS-232 and TTL inputs for external control and status feedback to a computer, if desired. The pumps can be daisy chained for multi-pump applications involving push/pull protocols, or multiple independent channels.
  • Cost-effective
  • Rich command set
  • RS-232 and TTL control
  • AL-1000 Syringe Pump accepts plastic syringes up to 60 ml and selected glass micro syringes from 0.5 to 500 µl; AL-3000 holds syringes up to 60 ml
  • Infuse/Withdraw Pump (AL-1000) or Infuse only (AL-300)
  • Displays total volume dispensed in ml or µl units
  • Choose system: Standard AL-1000 (AL-1000), AL-1000 Pumps (AL-2000), or Infuse Only (AL-300)
Note:
AL-1000 is only available as single syringe capacity.
The AL-2000 is two AL-1000's for separate bench set-up. This is especially useful when inject/withdraw parameters are different for a multi-stage set-up. Syringe(s) not included.
Benefits:
Automatic dispensing of small volumes
Very precise, reproducible flow rate control
Hands-free operation with foot switch
Program sequences without a computer (AL-1000 series)
Selectable infusion rate units (ml/hr, µl/hr, ml/min, µl/min)
Applications:
Drug administration
Chemical applications with slow incorporation of fixed volumes of fluids
The Aladdin pump series arguably offers the best VALUE for any syringe pump on the market
These pumps are available in single, dual, 4, 6, and 8 syringe capacities. Although it is one of the most cost-effective pumps available, the Aladdin pump series boasts a versatility that is unmatched in its price range with features typically found only on pumps costing more than twice as much.
The Aladdin series provides the capability to both inject and withdraw. These pumps can be used for manually triggered injections, but also have the capability to be programmed in multi-step pumping sequences without the requirement for a computer. Even so, this series also has both RS-232 and TTL inputs for external control and status feedback to a computer, if desired. The pumps can be daisy chained for multi pump applications involving push/pull protocols or multiple independent channels. The performance characteristics are admirable, considering the price.
Dual Pumping Action: Daisy Chain AL-1000 Aladdins:
Two Aladdin pumps when daisy-chained are more efficient and affordable than any competitor's dual syringe models. Two AL-1000 Aladdins (AL-2000) will perform as a dual infusion/withdrawal pump, a double pump for infusing at different rates, a push/pull pump with one infusing and one withdrawing at the same or different rates, two independent pumps, or a master/slave pump. One Aladdin can even control the second for continuous pumping with optional check valve set. The Aladdin pump series will accept syringes from Becton Dickinson, Monoject, Terumo, and Air-Tite.