Wire Speculums
Supplier: WORLD PRECISION INSTRUMENTS LLC
14130 14130-G 500368
77661-782EA 48.68 CAD
77661-782 77661-784 77662-320
Wire Speculums
Specula
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye.
  • Small, wire eyelid speculum 5 cm long
  • 10×5 mm blades
  • 5 mm depth
  • Stainless steel or premium German steel
  • Autoclavable
Our small wire speculum is made of surgical grade stainless steel or our premium German steel, and it comes in two sizes, with a spread of 13 and 25 mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.