Human Recombinant EGF (CTGrade GMP) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Fibroblast growth factor 8 (FGF-8) is a critical mitogenic factor that is required for normal development of the eye, ear, brain, and limb. FGF-8 functions broadly to promote cell proliferation, differentiation, and migration. Overexpression of FGF-8 increases tumor growth and angiogenesis. Human and mouse FGF-8 proteins show 100% homology.
- High quality
- Low endotoxin
- Lot-to-lot consistency
- Animal free
- Manufactured following GMP guidelines
Upon receipt, store the lyophilized protein at -20°C in a manual defrost freezer. Unopened vials are stable for 36 months from the date of manufacture. Reconstituted material should be apportioned in working volumes and stored at or below –20 °C in manual defrost freezer. For short term storage reconstituted material is stable for 4 to 6 weeks when stored at 2 to 8 °C. Stability can be increased by adding at least 0.1% carrier protein.
Host cell protein: ≤30 ng/ml
Host cell DNA: ≤0.01 ng/ml
Sterility testing: No growth
Mycoplasma: Negative
Caution: For research use only.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...