Vessel Clips
Supplier: WORLD PRECISION INSTRUMENTS LLC
501779-G 501780-G 501784-G 501788-G
77662-852EA 156.42 CAD
77662-852 77662-854 77662-856 77662-858
Vessel Clips
First Aid Supplies Wound Clips
Vessel clips are a surgical instrument for use in microvascular procedures, where extremely small blood vessels and delicate tissues are involved.
  • Perfect for microsurgery for hemostasis, occluding small blood vessels
  • Straight tips
  • Autoclavable and reusable
  • German steel
Vessel clips are significantly smaller than standard surgical clips, because they are designed to occlude fine blood vessels with precise control. These ligating vessel clips can tie off blood vessels or other tissues. Micro vessel clips are made to be durable and withstand theMore Product Information