Microsurgical Knives
Supplier: WORLD PRECISION INSTRUMENTS LLC
500249 500250 501731
77662-240PK 223.85 CAD
77662-240 77662-242 77662-746
Microsurgical Knives
Knives
This microsurgical knife is a disposable surgical instrument used in microsurgery on very small structures, often under a microscope.
  • Thin, fine blade with a 15° angle
  • Not autoclavable
  • Disposable knife for microdissection
Microsurgical knives have an extremely fine and sharp blade, allowing for precise and controlled incisions in delicate tissues. These knives have a straight blade that is super thin and narrow, allowing you to make precise cuts with minimal damage to surrounding tissues. These disposable blades are often used for ophthalmic surgery, neurosurgery, vascular surgery, reconstructive surgery, plastic surgery, and microdissection.
These disposable microsurgical knives have a knurled plastic handle that makes the knives lightweight and offers a secure hold. These are designed as disposable surgical knives.
Packaging: Individually wrapped and sold in packages of six.