KIMAX® Schellbach Burets, Class B, Straight Bore PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17150F-50 17150F-25
17454-727CS 353.51 CAD
17454-727 17454-705
KIMAX® Schellbach Burets, Class B, Straight Bore PTFE Stopcock, Kimble Chase
Burettes
Schellbach burets have a white band with a vertical blue stripe behind the black enamel scale to optically enhance the meniscus for increased accuracy
Delivery is facilitated by the straight bore PTFE stopcock.
Ordering information: Burets are shipped complete with buret body, stopcock and KIM-KAP® dust cap.