KIMAX® Burets, Serialized and Certified, Class A, Three-Way PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17052F-25 17052F-100 17052F-50
17527-081CS 578.53 CAD
17527-081 17527-128 17527-106
KIMAX® Burets, Serialized and Certified, Class A, Three-Way PTFE Stopcock, Kimble Chase
Burettes
Each KIMAX® precision bore buret has a permanent unique serial number and durable white enameled scale
Manufactured according to ASTM E287 Class A requirements.
Fill through the stopcock using 0.25" I.D. flexible tubing. Stopcock [PS] 2.
Ordering information: Shipped complete with KIM-KAP® dust cap and Certificate of Graduation Accuracy. Shipped in NSTA approved packaging.