KIMAX® Schellbach Burets, Automatic Self-Zeroing, Three-Way PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17170F-25 17170F-50
17552-158CS 737.74 CAD
17552-158 17552-191
KIMAX® Schellbach Burets, Automatic Self-Zeroing, Three-Way PTFE Stopcock, Kimble Chase
Burettes
KIMAX® borosilicate glass with a white Schellbach band and a vertical blue stripe behind a black enamel scale for repeatable accuracy
Buret is designed to be self-zeroing, with 0.25" O.D. overflow and filling tubes and a three-way PTFE stopcock. Calibrated to be read at the apparent meniscus. Use of the “true” meniscus may result in a lesser volume being dispensed from zero.
Ordering information: Burets are shipped complete with PTFE stopcock.