KIMAX® Burets, Automatic Self-Zeroing, Class B, Three-Way PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17051F-10 17051F-25 17051F-50 17051F-100
17550-061CS 656.59 CAD
17550-061 17550-083 17550-108 17550-120
KIMAX® Burets, Automatic Self-Zeroing, Class B, Three-Way PTFE Stopcock, Kimble Chase
Burettes
Reliable KIMAX® borosilicate construction with durable black enamel graduations
Manufactured according to ASTM E287 Class B requirements.
Side fill and overflow both accept 1/4" flexible tubing. Filling is through stopcock. Calibrated from the tip of the overflow tube to deliver volume indicated.
Ordering information: Shipped in NSTA approved packaging.