KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
Supplier: DWK Life Sciences (KIMBLE)
17054F-100 17054F-25 17054F-50 17054F-10
17456-188CS 949.59 CAD
17456-188 89001-634 89001-636 17456-182
KIMAX® Reservoir Buret, Automatic Self-Zeroing, Serialized
Burettes
Class-A burettes ideal for repeat titrations requiring traceable volumetric accuracy.
  • Self-zeroing
  • Easy-to-read durable black enamel scale
  • Permanently marked with an individual serial number and traceable to NIST standards
  • Manufactured from 33 expansion, low extractable borosilicate glass
Filling and the overflow tubes accept 1/4" I.D. flexible tubing.
Certifications: Designed from ASTM E287, Class A requirements. Manufactured to the specifications found in ASTM E694. Calibrated to the accuracy requirements found in ASTM E542.
Delivery information: Supplied with a Certificate of Graduation Accuracy.