Vannas Tübingen Scissors, Ceramic Coated
Supplier: WORLD PRECISION INSTRUMENTS LLC
504499 504488
77663-640EA 579.05 CAD
77663-640 77663-622
Vannas Tübingen Scissors, Ceramic Coated
Scissors
Ceramic coated Vannas Tübingen scissors are specialized microsurgical scissors that are highly precise and delicate, commonly used in ophthalmic and neurological procedures.
  • Ambidextrous spring scissors for delicate surgeries
  • Non-reflective, great for microscopy work or use under surgical lights
  • Durable coating for scissors that stay sharp longer
  • Curved or straight blades
  • Double spring thumb scissors
  • Surgical grade stainless steel with black ceramic coating
  • German manufactured
The black coating is non-reflective. Vannas Tübingen scissors are incredibly fine instruments, crafted of high-quality surgical grade stainless steel with extreme precision. Their design includes thin, and sharp blades, which allow for highly accurate and precise cutting in microsurgical settings. The tips of these scissors are exceptionally sharp and thin, enabling them to access and cut very fine and delicate tissues without causing unnecessary damage to surrounding structures. This is especially crucial in delicate surgeries, such as in ophthalmic procedures, where precision is paramount.
The blades of Vannas Tübingen scissors can be curved or straight. This curvature allows for better access and visibility in tight spaces or when working around delicate structures. These spring scissors are ambidextrous by nature, often preferred by left handed people. Like most surgical instruments, Vannas Tübingen scissors are designed to be sterilizable, allowing for safe and hygienic use in surgical settings. The combination of their ultra-fine, sharp tips, curved design, and exceptional precision makes Vannas Tübingen scissors essential tools in microsurgical settings, particularly in delicate eye surgeries and neurological procedures where the utmost precision and care are required to ensure successful outcomes with minimal damage to surrounding tissues.