Multidrop® Combi nL Reagent Dispenser, Thermo Scientific
Supplier: Thermo Scientific
Multidrop™
5840400 5840400
CA95039-854EA 168120.3 CAD
CA95039-854 95039-854
Multidrop® Combi nL Reagent Dispenser, Thermo Scientific
Microplate Dispensers
Nano- to micro-volume bulk reagent dispenser features precise, accurate, and reliable submicroliter dispensing for day-to-day reproducibility and high-quality assay data in pharmaceutical and biotechnology laboratories
The fast dispensing speed and full robot compatibility provides increased assay throughput for laboratories needing low-volume assay formats. Intuitive visual user interface ensures that all functions are easy to set up and use. Additionally, the instrument can be controlled with easy-to-use PC software. Serial RS-232 and USB interface connections. Shaker function included. Compatible plate types: 96-, 384-, and 1535-well plates.
Ordering information: Factory developed installation, operation, and performance qualification document is approved by design engineers and written in accordance with standards and guidelines. The qualification services are carried out by certified technicians on site.