Sapphire Scalpel Blades
Supplier: WORLD PRECISION INSTRUMENTS LLC
504168 504072 504073 504074 504075 504076 504077
77663-520EA 313.88 CAD
77663-520 77663-480 77663-482 77663-484 77663-486 77663-488 77663-490
Sapphire Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Sapphire Blades are a precise and durable solution for use in microsurgery, dissection, and related applications. Sapphire blades are created using a chemical process which forms an artificial sapphire.
  • Sapphire blades for surgical applications
  • 0.20 mm thick
  • Variety of shapes
The sapphire has a hardness value of 9 on the Mohs hardness scale, slightly below the hardest mineral on earth, the diamond. This characteristic provides supreme durability and sharpness to the blade. The sapphire blade's ultra sharp edge creates fine and precise cuts that prevent tissue damage and promote faster healing with less scarring.
The sapphire knife's ergonomic grip offers an easier grasp and is designed to be used with WPI's blades. The blade's chemical composition provides a durable edge that lasts longer than steel surgical instruments, saving costs for your practice.
The blade's sharp point and transparent finish allows greater precision in hair restoration, ophthalmic microsurgery, or cosmetic procedures. Sapphire blades may be sterilized using any currently known procedure. The blades are corrosion free and resistant to saline solution.
Ordering information: Handles 77662-272 and 77662-862 are sold separately.