Needle Holders with Lock
Supplier: WORLD PRECISION INSTRUMENTS LLC
503354 503355
77663-276EA 304.15 CAD
77663-276 77663-278
Needle Holders with Lock
Forceps
Needle Holder with lock is a surgical instrument used for microsurgery requiring precise control and manipulation of small needles. These have a 0.4 mm rounded tip. Chose either straight or curved tips. The curved needle holders are a little longer.
Our Needle Holders feature fine tips for grasping and manipulation of small, delicate needles. These needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces.
They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. These Needle Holders are available in surgical grade stainless steel, and they are autoclavable.