Screw Cap Microtubes
Supplier: SARSTEDT INC
72.694.321 72.730.027 72.694.105 72.730.025 72.687 72.692.100
77588-812CS 612.17 CAD
77588-812 77588-944 77588-794 77588-940 77588-704 77588-770
Screw Cap Microtubes
Tubes Microtubes
SARSTEDT screw cap microtubes provide an exceptionally secure seal with the integrated O-ring. Ideal for centrifugation, transport and storage of high-quality oligonucleotides, enzymes and buffers.
  • Material: PP
  • Robust polypropylene material for safety and versatility
  • 3 different volumes meet filling requirements from a few microliters to 2 milliliters
  • Leak-resistant according to ADR and IATA- tested at a vacuum of 0.95 bar
  • Centrifugation up to 20000×g
  • Can be autoclaved to 121 °C and frozen to –80 °C
  • O-ring cap increases cap security (IATA tested)
With volumes from 0.5 to 2.0 ml, a variety of container styles and caps, plus a wide selection of coding options, SARSTEDT screw cap micro tubes offer the right solution for your every requirement.