Cell Culture Chambers, x-well PCA

Supplier: SARSTEDT INC
94.6140.102 94.6140.202 94.6140.402
77589-854CS 1655.59 CAD
77589-854 77589-856 77589-858
Cell Culture Chambers, x-well PCA
Slides Chamber Slides
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.

  • Design: On PCA slide
  • Removable frame: Yes
  • Cell type: Adherent cells
  • Material: PCA
  • Frame material: PS
  • Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Cell culture chambers for the analysis of adherent cell growth on slides in the standard format
  • Single and multiple-chamber vessels for fluorescence or light microscope analyses of live or fixated cells
  • Small compartments for conducting individual examinations or parallel test series in a cost-effective and time-efficient way
  • PCA (polyolefin plastic) slides have a lower autofluorescence and a higher chemical resistance than polystyrene
  • Chamber can be released from the slide without any tools

The x-well cell culture system is available in various slide material options for a multitude of microscopic cell culture analyses. PCA, glass, coverglass or lumox® film slides combined with a polystyrene frame form one- and multiple-chamber vessels. x-well cell culture chambers make parallel tests quick and easy.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR