Low Retention Pipette Tips
Supplier: SARSTEDT INC
70.3031.110 70.3031.210 70.3020.210 70.3020.275 70.3030.110 70.3040.375 70.3040.110 70.3040.210 70.3040.275 70.3010.110 70.3030.210 70.3030.275 70.3030.375 70.3030.385 70.3050.375 70.3060.110 70.3060.210 70.3050.110 70.3050.210 70.3060.275
77588-506CS 260.26 CAD
77588-506 77588-512 77588-458 77588-462 77588-476 77588-544 77588-528 77588-534 77588-538 77588-422 77588-482 77588-488 77588-498 77588-500 77588-568 77588-574 77588-580 77588-552 77588-558 77588-584
Low Retention Pipette Tips
Pipette Tips
With our proven, high-quality tips you can expect maximum precision, sophisticated design and the highest purity. By using our low retention tips, costly and sensitive reagents can be more efficiently transferred with less loss, improving test results.
  • One system with 4 ordering options to fit your every requirement: Box, SingleRefill, StackPack and Bag
  • The SingleRefill uses 41% and the StackPack uses 64% less material than continually ordering a regular box system
  • Reduces waste - after use, the individual components of the box system can fit into each other. This saves disposal volume in turn reducing the waste produced
  • Volume, batch and shelf life are laser coated on each tray for easy identification
The Box - Universal and robust solution at the heart of our pipette tip system. The SingleRefill - Easy way to refill filter tips and eliminate unnecessary waste by reusing existing box. The closed design prevents contamination of the pipette tips, meaning the refill process is hygienic as well as environmentally friendly. The StackPack - Higher sample processing rates require more efficient solutions. Individual trays can be transferred to the box in one simple maneuver, plus the compact and stable design means that tips can be removed for use directly from the StackPack itself. The Bag - Provides a low-cost packaging solution, which can be hygienically opened and sealed again using the pressure seal. Additional tamper-evident seal also gives users that extra reassurance that tips are 100% free from contamination.