Disposal Bag, PP, Transparent, with Pressure
Supplier: SARSTEDT INC
86.1202 86.1203
77589-540CS 365.17 CAD
77589-540 77589-542
Disposal Bag, PP, Transparent, with Pressure
Bags Waste Disposal Bags
SARSTEDT offers a range of disposal bags for autoclaving and disposal of contaminated laboratory products.
  • Print: With print
  • For disposal of contaminated products from laboratories and hospitals
  • Strong, 50 µm thick PP film for high tear resistance
  • Suitable for steam sterilization in autoclaves at 134 °C
  • Always autoclave disposal bag unsealed
  • Please dispose of sharp objects in our 'disposal boxes' product range
Ideal for the containment and disposal of contaminated laboratory and hospitable disposables.