Inoculation Needles
Supplier: SARSTEDT INC
86.1568.010 86.1568.050
77589-578CS 233.51 CAD
77589-578 77589-580
Inoculation Needles
Spreaders Inoculating Loops and Needles
One aspect of microbiology is researching and identifying pathogens, as well as looking for suitable treatments. SARSTEDT provides researchers with the consumables they need to do their work.
  • Material: PS
  • Ideal for transferring very small viable microorganisms (inocula)
  • Also suitable for inoculating stab cultures
  • Saves time when processing larger test series
  • Safe handling - no pathogenic aerosols or cross contamination
  • Sterilized by gamma irradiation
Flexible SARSTEDT inoculation loops allow swabs to be taken easily or inoculation in liquid media. The inoculation needle can be used to inoculate or remove individual colonies. We recommend use of the inoculation spreader to evenly apply larger volumes to culture
media.