Vannas Spring Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
501777 501930
77662-848EA 218.99 CAD
77662-848 77662-956
Vannas Spring Scissors
Scissors
Our Vannas spring scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers.
  • Perfect for ambidextrous use, prefered by left hand users
  • 500 µm extra fine tips
  • Angled, straight or curved tips
  • Titanium or surgical grade stainless steel
Compared with other spring scissors, Vannas spring scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Like all spring scissors, they are perfect for ambidextrous use and are often preferred by left-handed people. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion.
Our titanium scissors are lightweight and non-magnetic. These Vannas have a fine 500 µm tip and 8 mm blades, making them suitable for surgery on small animals or researchers working with delicate tissues. They are autoclavable. Like all fine surgical instruments, these Vannas surgical scissors should be cleaned promptly after every use.