Iris Forceps, Serrated Tips
Supplier: WORLD PRECISION INSTRUMENTS LLC
15914 15914-G 15915 15915-G
77661-926EA 90.03 CAD
77661-926 77661-928 77661-930 77661-932
Iris Forceps, Serrated Tips
Forceps
Iris forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma. The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. Researchers like these fine tipped tweezers for precision work in microdissection.
  • Iris forceps for fine, precision work
  • 10 cm (4") Long
  • 0.8 mm Fine tips
  • Serrated jaws
  • Straight or curved tips
  • Surgical grade stainless steel or premium German steel
The Iris forceps are manufactured with surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.